beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
Thermo Scientific Chemicals
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8 | CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA410995000
Price (MYR)
2,562.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,562.00
EA
Add to cart
Specifications
Chemical Name or Materialβ-Glycerophosphoric acid, disodium salt pentahydrate
CAS13408-09-8
CAS Min %97.5
Health Hazard 1
CAS Max %100.0