Glycerol, 99.6%, ACS reagent
Glycerol, 99.6%, ACS reagent
Glycerol, 99.6%, ACS reagent
Glycerol, 99.6%, ACS reagent
Thermo Scientific Chemicals
Glycerol, 99.6%, ACS reagent
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA410985000
Price (MYR)
530.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
530.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
Health Hazard 1HYGROSCOPIC
Decomposition Information>290°C
Melting Point18.0°C