Glutaric dialdehyde, 8% solution in water
Glutaric dialdehyde, 8% solution in water
Glutaric dialdehyde, 8% solution in water
Glutaric dialdehyde, 8% solution in water
Thermo Scientific Chemicals
Glutaric dialdehyde, 8% solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Catalog NumberQuantity
FSA410971000100 g
Catalog number FSA410971000
Price (MYR)
588.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
588.00
BOT
Add to cart
Chemical Identifiers
CAS111-30-8, 7732-18-5
Specifications
Chemical Name or MaterialGlutaric dialdehyde
Physical FormSolution
CAS111-30-8
Density1.0160g/mL
Health Hazard 1Danger