Glutaric dialdehyde, 50 wt.% solution in water
Glutaric dialdehyde, 50 wt.% solution in water
Glutaric dialdehyde, 50 wt.% solution in water
Glutaric dialdehyde, 50 wt.% solution in water
Thermo Scientific Chemicals
Glutaric dialdehyde, 50 wt.% solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA410960010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-30-8, 7732-18-5
Specifications
Chemical Name or MaterialGlutaric dialdehyde
Name Note50 wt% Solution in Water
CAS111-30-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Very toxic