Geraniol, 99%
Geraniol, 99%
Geraniol, 99%
Thermo Scientific Chemicals
Geraniol, 99%
CAS: 106-24-1 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA410905000
Price (MYR)
1,509.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,509.00
EA
Add to cart
Specifications
Chemical Name or MaterialGeraniol
CAS106-24-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for