Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Thermo Scientific Chemicals
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA410730050
Price (MYR)
1,146.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,146.00
EA
Add to cart
Chemical Identifiers
CAS50-00-0, 7732-18-5, 67-56-1
Specifications
Chemical Name or MaterialFormaldehyde
Name Note37 wt% Solution
CAS50-00-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes damage to organs.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause cancer.
Fl
View more
General Description
  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol
Applications
  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
RUO – Research Use Only
General References:
  1. Ramanathan, M.; Porter, D. F.; Khavari P. A. Methods to study RNA-protein interactions. Nat Methods. 2019, 16(3), 225-234. Erratum in: Nat Methods. 2019, 16(4), 351.