2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
Thermo Scientific Chemicals
2-Ethylhexanal, 96%
CAS: 123-05-7 | C8H16O | 128.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA410092500
Price (MYR)
406.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
406.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Ethylhexanal
Name Notepract., 99%
Acidity8mg KOH/g max.
CAS123-05-7
CAS Min %95.0