Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA409930010
Price (MYR)
717.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
717.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid
CAS60-00-4
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Decomposition Information>150°C
Health Hazard 3GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.