Dipropylene glycol, 99%, mixture of isomers
Dipropylene glycol, 99%, mixture of isomers
Dipropylene glycol, 99%, mixture of isomers
Dipropylene glycol, 99%, mixture of isomers
Thermo Scientific Chemicals
Dipropylene glycol, 99%, mixture of isomers
CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA409150030
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
3 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialDipropylene glycol
Name Note99%
CAS25265-71-8
ColorOrange-Red to Yellow
Density1.0200g/mL
View more
Note: The chemical structure above shows only one of the molecules included in this mixture.
RUO – Research Use Only