1,4-Dinitrobenzene, 98%
1,4-Dinitrobenzene, 98%
1,4-Dinitrobenzene, 98%
1,4-Dinitrobenzene, 98%
Thermo Scientific Chemicals
1,4-Dinitrobenzene, 98%
CAS: 100-25-4 | C6H4N2O4 | 168.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA408650250
Price (MYR)
1,364.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,364.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 4-Dinitrobenzene
CAS100-25-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0