Dimethylthiocarbamoyl chloride, 99%
Dimethylthiocarbamoyl chloride, 99%
Dimethylthiocarbamoyl chloride, 99%
Dimethylthiocarbamoyl chloride, 99%
Thermo Scientific Chemicals
Dimethylthiocarbamoyl chloride, 99%
CAS: 16420-13-6 | C3H6ClNS | 123.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA408610250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethylthiocarbamoyl chloride
Name Note99%
CAS16420-13-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
Reacts violently with water.