Dimethylglyoxime, disodium salt octahydrate, 99%
Dimethylglyoxime, disodium salt octahydrate, 99%
Dimethylglyoxime, disodium salt octahydrate, 99%
Dimethylglyoxime, disodium salt octahydrate, 99%
Thermo Scientific Chemicals
Dimethylglyoxime, disodium salt octahydrate, 99%
Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19 | CAS: 75006-64-3 | C4H22N2Na2O10 | 304.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA408342500
Price (MYR)
1,046.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,046.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethylglyoxime, disodium salt octahydrate
Name Note99%
CAS75006-64-3
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.