N,N-Dimethyldodecylamine, 95%, pract.
N,N-Dimethyldodecylamine, 95%, pract.
N,N-Dimethyldodecylamine, 95%, pract.
N,N-Dimethyldodecylamine, 95%, pract.
Thermo Scientific Chemicals
N,N-Dimethyldodecylamine, 95%, pract.
CAS: 112-18-5 | C14H31N | 213.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA408292500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethyldodecylamine
Name Notepract., 95%
CAS112-18-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger