7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
7,12-Dimethylbenz[a]anthracene, 98%
Thermo Scientific Chemicals
7,12-Dimethylbenz[a]anthracene, 98%
CAS: 57-97-6 | C20H16 | 256.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA408181000
Price (MYR)
523.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
523.00
BOT
Add to cart
Specifications
Chemical Name or Material7, 12-Dimethylbenz[a]anthracene
Name Note99%
CAS57-97-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger