3,3'-Dimethoxybenzidine, 97%
3,3'-Dimethoxybenzidine, 97%
3,3'-Dimethoxybenzidine, 97%
3,3'-Dimethoxybenzidine, 97%
Thermo Scientific Chemicals
3,3'-Dimethoxybenzidine, 97%
CAS: 119-90-4 | C14H16N2O2 | 244.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA407890050
Price (MYR)
383.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
383.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 3'-Dimethoxybenzidine
Name Notepract., 97%
CAS119-90-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger