N,N-Diethyl-p-phenylenediamine oxalate, 96%
N,N-Diethyl-p-phenylenediamine oxalate, 96%
N,N-Diethyl-p-phenylenediamine oxalate, 96%
N,N-Diethyl-p-phenylenediamine oxalate, 96%
Thermo Scientific Chemicals
N,N-Diethyl-p-phenylenediamine oxalate, 96%
CAS: 62637-92-7 | 0·5 C2H2O4 | 209.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA407410250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Diethyl-p-phenylenediamine oxalate
Name Note96%
CAS62637-92-7
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning