N,N-Diethyl-p-phenylenediamine, 98%
N,N-Diethyl-p-phenylenediamine, 98%
N,N-Diethyl-p-phenylenediamine, 98%
N,N-Diethyl-p-phenylenediamine, 98%
Thermo Scientific Chemicals
N,N-Diethyl-p-phenylenediamine, 98%
CAS: 93-05-0 | C10H16N2 | 164.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA407391000
Price (MYR)
1,112.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,112.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethyl-p-phenylenediamine
CAS93-05-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0