Diethyl oxalacetate sodium salt, 95%, pract.
Diethyl oxalacetate sodium salt, 95%, pract.
Diethyl oxalacetate sodium salt, 95%, pract.
Diethyl oxalacetate sodium salt, 95%, pract.
Thermo Scientific Chemicals
Diethyl oxalacetate sodium salt, 95%, pract.
CAS: 40876-98-0 | C8H11NaO5 | 210.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA407375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl oxalacetate sodium salt
CAS40876-98-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.