Diethyl iminodiacetate, 98%
Diethyl iminodiacetate, 98%
Diethyl iminodiacetate, 98%
Diethyl iminodiacetate, 98%
Thermo Scientific Chemicals
Diethyl iminodiacetate, 98%
CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA407320050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl iminodiacetate
Name Note99%
CAS6290-05-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: