Diethyl adipate, 99%
Diethyl adipate, 99%
Diethyl adipate, 99%
Diethyl adipate, 99%
Thermo Scientific Chemicals
Diethyl adipate, 99%
CAS: 141-28-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA407175000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl adipate
CAS141-28-6
Melting Point-18.0°C
ColorColorless
Density1.0000g/mL