Didecyl phthalate, 95%
Didecyl phthalate, 95%
Didecyl phthalate, 95%
Didecyl phthalate, 95%
Thermo Scientific Chemicals
Didecyl phthalate, 95%
CAS: 84-77-5 | C28H46O4 | 446.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA407101000
Price (MYR)
1,970.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,970.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDidecyl phthalate
Name Note95%
CAS84-77-5
Melting Point4.0°C
Density0.9700g/mL