trans-1,2-Dichloroethylene, 99+%, stabilized
trans-1,2-Dichloroethylene, 99+%, stabilized
trans-1,2-Dichloroethylene, 99+%, stabilized
trans-1,2-Dichloroethylene, 99+%, stabilized
Thermo Scientific Chemicals
trans-1,2-Dichloroethylene, 99+%, stabilized
CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number FSA406842500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-1, 2-Dichloroethylene
Name Note99.7%
CAS156-60-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.