Dibutyl sebacate, 93%
Dibutyl sebacate, 93%
Dibutyl sebacate, 93%
Dibutyl sebacate, 93%
Thermo Scientific Chemicals
Dibutyl sebacate, 93%
CAS: 109-43-3 | C18H34O4 | 314.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA406652500
Price (MYR)
299.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
299.00
EA
Add to cart
Specifications
Chemical Name or MaterialDibutyl sebacate
CAS109-43-3
Melting Point-12°C
Density0.9400g/mL
EINECS Number203-672-5