Dibutyl phosphate, 97%
Dibutyl phosphate, 97%
Dibutyl phosphate, 97%
Dibutyl phosphate, 97%
Thermo Scientific Chemicals
Dibutyl phosphate, 97%
CAS: 107-66-4 | C8H19O4P | 210.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Glass bottle
Catalog number FSA406645000
Price (MYR)
585.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
585.00
EA
Add to cart
Specifications
Chemical Name or MaterialDibutyl phosphate
Name Note97%
Acidity259 to 267mg KOH/g
CAS107-66-4
Health Hazard 1GHS Signal Word: Danger