trans-1,4-Diaminocyclohexane, 98+%
trans-1,4-Diaminocyclohexane, 98+%
trans-1,4-Diaminocyclohexane, 98+%
trans-1,4-Diaminocyclohexane, 98+%
Thermo Scientific Chemicals
trans-1,4-Diaminocyclohexane, 98+%
CAS: 2615-25-0 | C6H14N2 | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA406360250
Price (MYR)
844.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
844.00
BOT
Add to cart
Specifications
Chemical Name or Materialtrans-1, 4-Diaminocyclohexane
CAS2615-25-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat