Dextran, for biochemistry, low fraction
Dextran, for biochemistry, low fraction
Dextran, for biochemistry, low fraction
Dextran, for biochemistry, low fraction
Thermo Scientific Chemicals
Dextran, for biochemistry, low fraction
CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA406271000
Price (MYR)
897.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
897.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDextran
ColorWhite
Name NoteLow fraction, For biochemistry
pH4.5 to 7.0 (6% aq. soln.)
CAS9004-54-0