L-Cystine, specified according to the requirements of Ph.Eur.
CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol
Have Questions?
Change view
Quantity:
100 g
25 g
Catalog number FSA406161000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Cystine
Melting Point260.0°C to 261.0°C
Ammonium200ppm max.
CAS56-89-3
Heavy Metals (as Pb)10ppm max.