beta-Cyclodextrin, 98%
beta-Cyclodextrin, 98%
beta-Cyclodextrin, 98%
beta-Cyclodextrin, 98%
Thermo Scientific Chemicals
beta-Cyclodextrin, 98%
CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA406005000
Price (MYR)
2,229.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,229.00
EA
Add to cart
Specifications
Chemical Name or Materialβ-Cyclodextrin
CAS7585-39-9
Melting Point310.0°C
EINECS Number231-493-2
Loss on Drying14% max. (105°C, 8 hrs)