Cresyl Violet acetate, pure, certified
Cresyl Violet acetate, pure, certified
Cresyl Violet acetate, pure, certified
Cresyl Violet acetate, pure, certified
Thermo Scientific Chemicals
Cresyl Violet acetate, pure, certified
CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA405760100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCresyl Violet acetate
Melting Point140.0°C to 143.0°C
CAS10510-54-0
Absorption Ratio0.94 to 1.2
Health Hazard 2GHS H Statement: