p-Cresol, 99+%, pure
p-Cresol, 99+%, pure
p-Cresol, 99+%, pure
p-Cresol, 99+%, pure
Thermo Scientific Chemicals
p-Cresol, 99+%, pure
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA405745000
Price (MYR)
528.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
528.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Cresol
Name Note99+%
CAS106-44-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.