Cobaltocene, 98%
Cobaltocene, 98%
Cobaltocene, 98%
Thermo Scientific Chemicals
Cobaltocene, 98%
CAS: 1277-43-6 | C10H10Co | 189.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA40532010010 gGlass bottle
Catalog number FSA405320100
Price (MYR)
2,810.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,810.00
EA
Add to cart
Specifications
Chemical Name or MaterialCobaltocene
CAS1277-43-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable solid.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.