Benzyl chloride, 99%, pure, stabilized
Benzyl chloride, 99%, pure, stabilized
Benzyl chloride, 99%, pure, stabilized
Benzyl chloride, 99%, pure, stabilized
Thermo Scientific Chemicals
Benzyl chloride, 99%, pure, stabilized
CAS: 100-44-7 | C7H7Cl | 126.583 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Catalog number FSA405092500
Price (MYR)
270.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
270.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyl chloride
Name NoteStabilized, Pure
CAS100-44-7
CAS Min %98.5
Health Hazard 1Danger
View more
Alkylation reagent
RUO – Research Use Only