3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
3-(Chlorosulfonyl)benzoyl chloride, 98%
Thermo Scientific Chemicals
3-(Chlorosulfonyl)benzoyl chloride, 98%
CAS: 4052-92-0 | C7H4Cl2O3S | 239.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA40507025025 gGlass bottle
Catalog number FSA405070250
Price (MYR)
1,604.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,604.00
BOT
Add to cart
Specifications
Chemical Name or Material3-(Chlorosulfonyl)benzoyl chloride
Name Note98%
CAS4052-92-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning