5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
Thermo Scientific Chemicals
5-Chlorosalicylaldehyde, 98%
CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA405030250
Price (MYR)
751.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
751.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Chlorosalicylaldehyde
Name Note98%
CAS635-93-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning