4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals
4-Chlorophenylhydrazine hydrochloride, 97%
CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA404950250
Price (MYR)
539.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
539.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chlorophenylhydrazine hydrochloride
Name Note97%
CAS1073-70-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning