2-Chloronaphthalene, 99%
2-Chloronaphthalene, 99%
2-Chloronaphthalene, 99%
2-Chloronaphthalene, 99%
Thermo Scientific Chemicals
2-Chloronaphthalene, 99%
CAS: 91-58-7 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA404840250
Price (MYR)
7,614.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,614.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chloronaphthalene
Name Note99%
CAS91-58-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning