1-Chlorohexane, 95%
1-Chlorohexane, 95%
1-Chlorohexane, 95%
Thermo Scientific Chemicals
1-Chlorohexane, 95%
CAS: 544-10-5 | C6H13Cl | 120.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA404645000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-chlorohexane
CAS544-10-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0