Cerium(IV) sulfate, 95%
Cerium(IV) sulfate, 95%
Cerium(IV) sulfate, 95%
Cerium(IV) sulfate, 95%
Thermo Scientific Chemicals
Cerium(IV) sulfate, 95%
CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA404341000
Price (MYR)
1,102.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,102.00
EA
Add to cart
Specifications
Chemical Name or MaterialCerium(IV) sulfate
Melting Point195.0°C
CAS13590-82-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.