Cellulose Acetate Butyrate, Butyryl Content 50-54%
Cellulose Acetate Butyrate, Butyryl Content 50-54%
Cellulose Acetate Butyrate, Butyryl Content 50-54%
Cellulose Acetate Butyrate, Butyryl Content 50-54%
Thermo Scientific Chemicals
Cellulose Acetate Butyrate, Butyryl Content 50-54%
CAS: 9004-36-8 | (C12H14O10)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA404225000
Price (MYR)
992.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
992.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCellulose acetate butyrate
ColorWhite
Name NoteButyryl Content 50-54%
CAS9004-36-8
Physical FormGranular Crystalline Powder