Castor oil
Castor oil
Castor oil
Thermo Scientific Chemicals
Castor oil
CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
2.5 kg
Catalog number FSA404165000
Price (MYR)
338.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
338.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCastor oil
CAS8001-79-4
Melting Point-10°C
ColorYellow
Density0.9570g/mL