5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
Thermo Scientific Chemicals
5(6)-Carboxyfluorescein
CAS: 72088-94-9 | C42H24O14 | 752.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number FSA404105000
Price (MYR)
309.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
309.00
BOT
Add to cart
Specifications
Chemical Name or Material5(6)-Carboxyfluorescein
Name Note99%
CAS72088-94-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement