Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Thermo Scientific Chemicals
Activated charcoal, NORIT™ CN 1, decolorizing
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Glass bottle
Catalog number FSA404045000
Price (MYR)
449.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
449.00
BOT
Add to cart
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* CN 1
Name NoteNORIT CN 1
CAS7440-44-0
Health Hazard 1GHS Signal Word: Warning
Melting Point3652.0°C
View more
Filter aid
RUO – Research Use Only