2-Butoxyethyl acetate, 98%
2-Butoxyethyl acetate, 98%
2-Butoxyethyl acetate, 98%
2-Butoxyethyl acetate, 98%
Thermo Scientific Chemicals
2-Butoxyethyl acetate, 98%
CAS: 112-07-2 | C8H16O3 | 160.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA403410010
Price (MYR)
341.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
341.00
EA
Add to cart
Specifications
Chemical Name or Material2-Butoxyethyl acetate
Name Note98%
CAS112-07-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning