Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol
Thermo Scientific Chemicals
Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3) | CAS: 373-57-9 | BF3 | 67.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA402761000
Price (MYR)
426.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
426.00
EA
Add to cart
Chemical Identifiers
CAS373-57-9, 67-56-1
Specifications
Boiling Point65.0°C
Chemical Name or MaterialBoron trifluoride
Melting Point-98.0°C
Name Note1.5M solution in methanol
CAS373-57-9
View more
Acylation reagent, Lewis acid
RUO – Research Use Only