Bis(2-ethylhexyl) adipate, 99%
Bis(2-ethylhexyl) adipate, 99%
Bis(2-ethylhexyl) adipate, 99%
Bis(2-ethylhexyl) adipate, 99%
Thermo Scientific Chemicals
Bis(2-ethylhexyl) adipate, 99%
CAS: 103-23-1 | C22H42O4 | 370.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA402462500
Price (MYR)
243.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
243.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBis(2-ethylhexyl) adipate
CAS103-23-1
CAS Min %98.5
Health Hazard 1
CAS Max %100.0