Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Benzenesulfonic acid, sodium salt, 98%
Thermo Scientific Chemicals
Benzenesulfonic acid, sodium salt, 98%
CAS: 515-42-4 | C6H5NaO3S | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA401851000
Price (MYR)
423.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
423.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzenesulfonic acid, sodium salt
Name Note98%
CAS515-42-4
CAS Min %97.5
Health Hazard 1HYGROSCOPIC