Barium diphenylaminesulfonate, pure, indicator grade
Barium diphenylaminesulfonate, pure, indicator grade
Barium diphenylaminesulfonate, pure, indicator grade
Barium diphenylaminesulfonate, pure, indicator grade
Thermo Scientific Chemicals
Barium diphenylaminesulfonate, pure, indicator grade
CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA401680500
Price (MYR)
2,641.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,641.00
EA
Add to cart
Specifications
Chemical Name or MaterialBarium diphenylaminesulfonate
CAS6211-24-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON
View more
Indicator
RUO – Research Use Only