L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur.
CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
Have Questions?
Change view
Quantity:
100 g
Catalog number FSA401481000
Price (MYR)
367.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
367.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Asparagine monohydrate
Melting Point233.0°C to 235.0°C
CAS5794-13-8
Molecular FormulaC4H10N2O4
Linear FormulaH2NCOCH2CH(NH2)CO2H·H2O