Anthrone, 95%, pure
Anthrone, 95%, pure
Anthrone, 95%, pure
Thermo Scientific Chemicals
Anthrone, 95%, pure
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA401361000
Price (MYR)
840.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
840.00
EA
Add to cart
Specifications
Chemical Name or MaterialAnthrone
CAS90-44-8
Health Hazard 1GHS Signal Word: Warning
Melting Point152.0°C to 157.0°C
EINECS Number201-994-