8-Anilino-1-naphthalenesulfonic acid, 98%
8-Anilino-1-naphthalenesulfonic acid, 98%
8-Anilino-1-naphthalenesulfonic acid, 98%
8-Anilino-1-naphthalenesulfonic acid, 98%
Thermo Scientific Chemicals
8-Anilino-1-naphthalenesulfonic acid, 98%
CAS: 82-76-8 | C16H13NO3S | 299.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA401211000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material8-Anilino-1-naphthalenesulfonic acid
CAS82-76-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0